Learn More
Kishigo P Series Mesh Vest
Catalog No. 19220980
Click to view available options
Color:
Orange
Catalog No. Color
19-220-980 Orange
1 options
This item is not returnable. View return policy
Catalog No. 19-220-980 Supplier Kishigo Supplier No. PV26
This item is not returnable. View return policy
Comfortable polyester mesh with cotton trim P Series Mesh Vest
  • Polyester mesh with cotton trim
  • 1 in. wide reflective stripes
  • 3 in. hook and loop front closure
  • 3/4 in. elastic side closures for universal sizing
Specifications
Closure Type Hook and Loop
Color Orange
Size One Size
Material Polyester Mesh