Learn More
Medchemexpress LLC P-Synephrine | 614-35-7 | 99.9% | 167.21 | 1 ML
SDP
Catalog No. 5000437001
Encompass_Preferred
p-Synephrine is an organic compound naturally occurring in various biofluids, including urine and blood. It is intended for research applications only.
  • Found in human biofluids
  • High purity for reliable research
  • Acts as a human endogenous metabolite
  • Derived from natural plant sources like Citrus x aurantium
  • Soluble in both DMSO and water for versatile experimental setups
Catalog No. 50-004-37001 Supplier Medchemexpress LLC Supplier No. HY11323610MM/1ML
p-Synephrine is an organic compound naturally occurring in various biofluids, including urine and blood. It is intended for research applications only.
  • Found in human biofluids
  • High purity for reliable research
  • Acts as a human endogenous metabolite
  • Derived from natural plant sources like Citrus x aurantium
  • Soluble in both DMSO and water for versatile experimental setups