Learn More
Taylor Chemical Drop Test, Alkalinity (P/T, for laundry), Phenolphthalein/Blended Indicator, 1 drop = 10 or 50 ppm
Catalog No. NC1578326
Encompass_Preferred
Alkalinity, P/T drop test using Phenolphthalein/blended indicator (laundry). 1 drop = 10 or 50 ppm P/T alkalinity as CaCO₃
Catalog No. NC1578326 Supplier Taylor Chemical Supplier No. K1512
Alkalinity, P/T drop test using Phenolphthalein/blended indicator (laundry). 1 drop = 10 or 50 ppm P/T alkalinity as CaCO₃