Learn More
Strem, An Ascensus Company CAS# 1621274-11-0. 250mg. Methanesulfonato(tri-t-butylphosphino)(2'-methylamino-1,1'-biphenyl-2-yl)palladium(II), 98% [P(t-Bu)3 Palladacycle Gen. 4]. MFCD28411657
SDP
Supplier:  Strem, An Ascensus Company 460385250MG
Encompass
CAS# 1621274-11-0. 250mg. Methanesulfonato(tri-t-butylphosphino)(2'-methylamino-1,1'-biphenyl-2-yl)palladium(II), 98% [P(t-Bu)3 Palladacycle Gen. 4]. MFCD28411657. Molecular Weight: 586.08. Molecular Formula: C26H42NO3PPdS. Color/form: tan to yellow pwdr. Strem# 46-0385. http://www.strem.com/catalog/v/46-0385/
Catalog No. 50-204-5361