Learn More
Bellco Glass, Inc. P/T Cult Tube,13x100mm
SDP
Catalog No. 501945288
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Non-corressive borosilicate glass to prevent etching. Accepts Bellco stainless steel closures and KAPUT closures. MADE IN THE USA.Specifications:Borosilicate GlassRound BottomSizes: 13x100mmVolume: 8.7ml’Case Quantity: 288
Catalog No. 50-194-5288 Supplier Bellco Glass, Inc. Supplier No. 201113100
Manufactured from Type 1 Class A Borosilicate Glass. Non-corressive borosilicate glass to prevent etching. Accepts Bellco stainless steel closures and KAPUT closures. MADE IN THE USA.Specifications:Borosilicate GlassRound BottomSizes: 13x100mmVolume: 8.7ml’Case Quantity: 288