Learn More
Bellco Glass, Inc. P/T Culture Tube,16x125mm Borosilicate Glass
SDP
Catalog No. 501945289
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Non-corressive borosilicate glass to prevent etching. Accepts Bellco stainless steel closures and KAPUT closures. MADE IN THE USA.Specifications:Borosilicate GlassRound BottomSizes: 16x125mmVolume: 17.4ml’Case Quantity: 144
Catalog No. 50-194-5289 Supplier Bellco Glass, Inc. Supplier No. 201116125
Manufactured from Type 1 Class A Borosilicate Glass. Non-corressive borosilicate glass to prevent etching. Accepts Bellco stainless steel closures and KAPUT closures. MADE IN THE USA.Specifications:Borosilicate GlassRound BottomSizes: 16x125mmVolume: 17.4ml’Case Quantity: 144