Learn More
Bellco Glass, Inc. P/T Culture Tube, 18x150mm
SDP
Catalog No. 501945290
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Non-corrosive Borosilicate glass to prevent etching. Accepts bellco stainless steel closures and kaput closures. MADE IN THE USA. Round bottom Sizes: 18x150mm Volume: 27.3ml Closure: 18mm
Catalog No. 50-194-5290 Supplier Bellco Glass, Inc. Supplier No. 201118150
Add to Cart
Add to Cart
Manufactured from type 1 class a Borosilicate glass. Non-corrosive Borosilicate glass to prevent etching. Accepts bellco stainless steel closures and kaput closures. MADE IN THE USA. Round bottom Sizes: 18x150mm Volume: 27.3ml Closure: 18mm