Learn More
Bellco Glass, Inc. P/T Culture Tube, 25x150mm
SDP
Catalog No. 501945291
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Non-corrosive Borosilicate glass to prevent etching. Accepts bellco stainless steel closures and kaput closures. MADE IN THE USA. Round bottom Sizes: 25x150mm Volume: 55.6ml Closure: 25mm
Catalog No. 50-194-5291 Supplier Bellco Glass, Inc. Supplier No. 201125150
Manufactured from type 1 class a Borosilicate glass. Non-corrosive Borosilicate glass to prevent etching. Accepts bellco stainless steel closures and kaput closures. MADE IN THE USA. Round bottom Sizes: 25x150mm Volume: 55.6ml Closure: 25mm