Learn More
Bellco Glass, Inc. Plain Top Leighton Tube,19X95mm Short Style
SDP
Catalog No. 501944674
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Extra long working area yet short overall length promotes convenient handling. Short neck allows easy entry and removal of cover slips. Also can fit bellco self-locking racks.MADE IN THE USA Tube dimensions: 19x95
Catalog No. 50-194-4674 Supplier Bellco Glass, Inc. Supplier No. 190319095
Manufactured from type 1 class a Borosilicate glass. Extra long working area yet short overall length promotes convenient handling. Short neck allows easy entry and removal of cover slips. Also can fit bellco self-locking racks.MADE IN THE USA Tube dimensions: 19x95