Learn More
Strem, An Ascensus Company CAS# 13716-12-6. 2g. Tri-t-butylphosphine, min. 98% (50wt% in toluene). MFCD00015006
SDP
Supplier:  Strem, An Ascensus Company 1558182G
Encompass
CAS# 13716-12-6. 2g. Tri-t-butylphosphine, min. 98% (50wt% in toluene). MFCD00015006. Molecular Weight: 202.32. Molecular Formula: C12H27P. Color/form: colorless liq. Strem# 15-5818. http://www.strem.com/catalog/v/15-5818/
Catalog No. 50-204-5256