Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich p-Terphenyl

Catalog No. 8647050G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
86-470-50G 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 86-470-50G Supplier Sigma Aldrich Supplier No. 8647050G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Valuable building block offered as a solution in ethanol for more convenient handling

Specifications

CAS 92-94-4
Melting Point 212°C to 213°C (lit.), 212°C to 214°C
Boiling Point 389°C (lit.)
Molecular Formula C18H14
Linear Formula C6H5C6H4C6H5
MDL Number MFCD00003061
Quantity 100 mL
Synonym 1,4-Diphenylbenzene; PT; PTP
Molecular Weight (g/mol) 230.3
Percent Purity ≥98.5% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.