Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Terphenyl, MilliporeSigma™ Supelco™

Catalog No. 111020006 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0006 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0006 Supplier MilliporeSigma Supelco Supplier No. 45802250MG
Add to Cart
Add to Cart
Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 92-94-4
Vapor Density 7.95 (vs. air)
Flash Point 207°C
Physical Form Neat
Linear Formula C6H5C6H4C6H5
MDL Number MFCD00003061
Melting Point 212°C to 213°C
Boiling Point 389°C
Synonym 1,4-Diphenylbenzene; PT; PTP
Type Analytical Standard
Quantity 250 mg
Formula Weight 230.30
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.