Learn More
Medchemexpress LLC P-Tolualdehyde | 104-87-0 | 99.10% | 120.15 | 500 G
SDP
Catalog No. 5000454783
Encompass_Preferred
p-Tolualdehyde is an active small molecule that functions as a drug intermediate. It is intended solely for research use and is not sold for patient treatment. This compound can be utilized in the synthesis of isoxazole derivatives, which show potential for developing multidrug resistance (MDR) reversal agents. Furthermore, p-Tolualdehyde serves as a model compound, reacting with the fluorescent probe DPSH to form DPSH-To, a more fluorescent compound suitable for aldehyde determination.
  • Used as a drug intermediate
  • Can be used for the synthesis of isoxazole derivatives
  • Potential for developing multidrug resistance (MDR) reversal agents
  • Model compound for aldehyde determination
  • Forms a more fluorescent compound (DPSH-To) with fluorescent probe DPSH
Catalog No. 50-004-54783 Supplier Medchemexpress LLC Supplier No. HYW012860500G
p-Tolualdehyde is an active small molecule that functions as a drug intermediate. It is intended solely for research use and is not sold for patient treatment. This compound can be utilized in the synthesis of isoxazole derivatives, which show potential for developing multidrug resistance (MDR) reversal agents. Furthermore, p-Tolualdehyde serves as a model compound, reacting with the fluorescent probe DPSH to form DPSH-To, a more fluorescent compound suitable for aldehyde determination.
  • Used as a drug intermediate
  • Can be used for the synthesis of isoxazole derivatives
  • Potential for developing multidrug resistance (MDR) reversal agents
  • Model compound for aldehyde determination
  • Forms a more fluorescent compound (DPSH-To) with fluorescent probe DPSH