Learn More
Medchemexpress LLC p-Tolualdehyde | 104-87-0 | 99.16% | 120.15 | 25 G
SDP
Catalog No. 5000451381
Encompass_Preferred
p-Tolualdehyde is an active small molecule primarily utilized as a drug intermediate. It appears as a colorless to light yellow liquid. This compound can be used for the synthesis of isoxazole derivatives, which are crucial for developing multidrug resistance reversal agents, and as a model compound for aldehyde determination using fluorescent probes.
  • Active small molecule
  • Used as a drug intermediate
  • Can be used for the synthesis of isoxazole derivatives
  • Functions as a model compound for aldehyde determination
  • Store at 4°C under nitrogen
  • Colorless to light yellow liquid appearance
Catalog No. 50-004-51381 Supplier Medchemexpress LLC Supplier No. 25GHYW012860
p-Tolualdehyde is an active small molecule primarily utilized as a drug intermediate. It appears as a colorless to light yellow liquid. This compound can be used for the synthesis of isoxazole derivatives, which are crucial for developing multidrug resistance reversal agents, and as a model compound for aldehyde determination using fluorescent probes.
  • Active small molecule
  • Used as a drug intermediate
  • Can be used for the synthesis of isoxazole derivatives
  • Functions as a model compound for aldehyde determination
  • Store at 4°C under nitrogen
  • Colorless to light yellow liquid appearance