Learn More
Medchemexpress LLC P-Tolualdehyde-d7 | 1219805-23-8 | 98.2% | 127.19 | 25 MG
SDP
Catalog No. 5000453196
Encompass_Preferred
P-Tolualdehyde-d7 is the deuterium-labeled p-Tolualdehyde. It is an active small molecule that can be used as a drug intermediate. This product is for research use only and is not sold to patients. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes into drug molecules for quantitation during drug development.
Catalog No. 50-004-53196 Supplier Medchemexpress LLC Supplier No. 25MGHYW012860S
P-Tolualdehyde-d7 is the deuterium-labeled p-Tolualdehyde. It is an active small molecule that can be used as a drug intermediate. This product is for research use only and is not sold to patients. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes into drug molecules for quantitation during drug development.