Learn More
Medchemexpress LLC p-Tolualdehyde-d7 | 1219805-23-8 | 98.2% | 127.19 | 5 MG
SDP
Catalog No. 5000454847
Encompass_Preferred
p-Tolualdehyde-d7 is the deuterium labeled p-Tolualdehyde, an active small molecule that can be used as a drug intermediate. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules primarily as tracers for quantitation during drug development. Deuteration has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs. This compound is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound.
Catalog No. 50-004-54847 Supplier Medchemexpress LLC Supplier No. HYW012860S5MG
p-Tolualdehyde-d7 is the deuterium labeled p-Tolualdehyde, an active small molecule that can be used as a drug intermediate. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules primarily as tracers for quantitation during drug development. Deuteration has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs. This compound is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound.