Learn More
Medchemexpress LLC p-Tolualdehyde-d7 | 1219805-23-8 | 98.2% | 127.19 | 50 MG
SDP
Catalog No. 5000454892
Encompass_Preferred
p-Tolualdehyde-d7 is the deuterium labeled p-Tolualdehyde. p-Tolualdehyde is an active small molecule that could be used as a drug intermediate. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-54892 Supplier Medchemexpress LLC Supplier No. HYW012860S50MG
p-Tolualdehyde-d7 is the deuterium labeled p-Tolualdehyde. p-Tolualdehyde is an active small molecule that could be used as a drug intermediate. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.