Learn More
AdipoGen p-Toluenesulfinic acid Na
SDP
Supplier:  AdipoGen CDXS0503G100
Encompass_Preferred
Chemical. CAS 824-79-3. Formula C7H7NaO2S. Molecular Weight 178.18. Sodium p-Toluenesulfinate is used as an organic bulding block or intermediate for the preparation of pharmaceuticals and dispersal dyes, as a mild reducing agent and as a polymerization accelerator. It is also used as a discharge medium of dyes and pigments. Used as a photo-redox couple together with methylene blue. Further, it serves as a chemical intermediate in the production of fluorescent pigments and fabric adhesive. In addition to this, it is used as an electroplating brightening agent and curing agent.
Catalog No. 50-256-0055