Learn More
Medchemexpress LLC P-toluenesulfonic acid-d7 monohydrate | 1219795-22-8 | 99.8% | 197.26 | C7H3D7O4S | 10 MG
SDP
Catalog No. 5000302644
Encompass_Preferred
p-Toluenesulfonic acid-d7 monohydrate is a deuterium-labeled analog of p-toluenesulfonic acid (monohydrate) used as a strong organic acid and catalyst in organic synthesis and analytical studies. The deuterium labeling supports mechanistic investigations and NMR tracing, and the material is supplied at high isotopic and chemical purity for research use.
  • Deuterium-labeled for tracing and NMR studies.
  • Monohydrate form for consistent handling and reproducibility.
  • High purity (99.76%) suitable for analytical and research applications.
  • Functions as a strong organic acid and catalyst in synthesis.
  • Available in small pack sizes for laboratory and analytical work.
Catalog No. 50-003-02644 Supplier Medchemexpress LLC Supplier No. HYW015175S10MG
p-Toluenesulfonic acid-d7 monohydrate is a deuterium-labeled analog of p-toluenesulfonic acid (monohydrate) used as a strong organic acid and catalyst in organic synthesis and analytical studies. The deuterium labeling supports mechanistic investigations and NMR tracing, and the material is supplied at high isotopic and chemical purity for research use.
  • Deuterium-labeled for tracing and NMR studies.
  • Monohydrate form for consistent handling and reproducibility.
  • High purity (99.76%) suitable for analytical and research applications.
  • Functions as a strong organic acid and catalyst in synthesis.
  • Available in small pack sizes for laboratory and analytical work.