Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC P-toluenesulfonic acid-d7 monohydrate | 1219795-22-8 | 99.8% | 197.26 | C7H3D7O4S | 5 MG
SDP

Catalog No. 5000302451
Encompass_Preferred

p-Toluenesulfonic acid-d7 monohydrate is a deuterium-labeled reagent supplied as a solid for research use. It is used as an acid catalyst and isotopic standard for mechanistic studies, mass spectrometry, and synthetic chemistry where deuterium labeling is required.

  • High purity (99.76%).
  • Chemical formula: C7H3D7O4S.
  • Molecular weight: 197.26 g·mol-1.
  • Appearance: white to off-white solid.
  • Available in small research quantities (e.g., 5 mg and 10 mg).
  • Store at -20 °C, protected from light, under inert atmosphere.
  • Useful as an isotopic tracer and internal standard in mass spectrometry.

Catalog No. 50-003-02451 Supplier Medchemexpress LLC Supplier No. HYW015175S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

p-Toluenesulfonic acid-d7 monohydrate is a deuterium-labeled reagent supplied as a solid for research use. It is used as an acid catalyst and isotopic standard for mechanistic studies, mass spectrometry, and synthetic chemistry where deuterium labeling is required.

  • High purity (99.76%).
  • Chemical formula: C7H3D7O4S.
  • Molecular weight: 197.26 g·mol-1.
  • Appearance: white to off-white solid.
  • Available in small research quantities (e.g., 5 mg and 10 mg).
  • Store at -20 °C, protected from light, under inert atmosphere.
  • Useful as an isotopic tracer and internal standard in mass spectrometry.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.