Learn More
Medchemexpress LLC P-toluenesulfonic acid-d7 monohydrate | 1219795-22-8 | 99.8% | 197.26 g/mol | C7H3D7O4S | 25 MG
SDP
Catalog No. 5000302218
Encompass_Preferred
p-Toluenesulfonic acid-d7 monohydrate is the deuterium-labeled monohydrate of p-toluenesulfonic acid. It functions as a strong organic acid and is used both as an acid catalyst in synthetic chemistry and as an internal standard or tracer for quantitative NMR, GC-MS, and LC-MS analyses.
  • Deuterium-labeled for use as an internal standard or tracer in NMR and mass spectrometry.
  • High isotopic purity (99.76%) for accurate quantitative analysis.
  • Solid form, easy to weigh and handle for small-scale analytical applications.
  • Suitable as an acid catalyst in organic synthesis.
  • Available in multiple small package sizes for analytical workflows.
Catalog No. 50-003-02218 Supplier Medchemexpress LLC Supplier No. 25MGHYW015175S
p-Toluenesulfonic acid-d7 monohydrate is the deuterium-labeled monohydrate of p-toluenesulfonic acid. It functions as a strong organic acid and is used both as an acid catalyst in synthetic chemistry and as an internal standard or tracer for quantitative NMR, GC-MS, and LC-MS analyses.
  • Deuterium-labeled for use as an internal standard or tracer in NMR and mass spectrometry.
  • High isotopic purity (99.76%) for accurate quantitative analysis.
  • Solid form, easy to weigh and handle for small-scale analytical applications.
  • Suitable as an acid catalyst in organic synthesis.
  • Available in multiple small package sizes for analytical workflows.