Learn More
Medchemexpress LLC p-toluenesulfonic acid-d7 monohydrate | 1219795-22-8 | 99.8% | 197.26 g/mol | C7H3D7O4S | 50 MG
SDP
Catalog No. 5000302657
Encompass_Preferred
p-Toluenesulfonic acid-d7 monohydrate is the deuterium-labeled monohydrate form of p-toluenesulfonic acid supplied for research use. It acts as a strong organic acid and catalyst in organic synthesis and is used as an isotopically labeled reagent or analytical standard for mass spectrometry and mechanistic studies.
  • Deuterium-labeled for isotopic tracing and analytical applications.
  • High chemical purity (99.76%).
  • Solid monohydrate form for convenient handling and storage.
  • Suitable as a strong acid catalyst in organic synthesis.
  • Available in small analytical pack sizes for research workflows.
Catalog No. 50-003-02657 Supplier Medchemexpress LLC Supplier No. HYW015175S50MG
p-Toluenesulfonic acid-d7 monohydrate is the deuterium-labeled monohydrate form of p-toluenesulfonic acid supplied for research use. It acts as a strong organic acid and catalyst in organic synthesis and is used as an isotopically labeled reagent or analytical standard for mass spectrometry and mechanistic studies.
  • Deuterium-labeled for isotopic tracing and analytical applications.
  • High chemical purity (99.76%).
  • Solid monohydrate form for convenient handling and storage.
  • Suitable as a strong acid catalyst in organic synthesis.
  • Available in small analytical pack sizes for research workflows.