Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich P-Toluenesulfonic Acid Monohydrate

Catalog No. T359205G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 kg
15 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T359205G 5 g
T35920500G 500 g
T3592015KG 15 kg
T35920100G 100 g
T359203KG 3 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. T359205G Supplier Sigma Aldrich Supplier No. T359205G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

P-Toluenesulfonic Acid Monohydrate

Specifications

CAS 6192-52-5
Melting Point 103°C to 106°C
Molecular Formula C7H8O3S -+ H2O
Linear Formula CH3C6H4SO3H -+ H2O
MDL Number MFCD00142137
Quantity 5 g
Molecular Weight (g/mol) 190.22 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.