Learn More
Medchemexpress LLC p-toluenesulfonic acid monohydrate | 6192-52-5 | MFCD00142137 | 99.8% | 190.22 g/mol | C7H10O4S | 1 ML
SDP
Catalog No. 5000303160
Encompass_Preferred
p-Toluenesulfonic acid monohydrate (CAS 6192-52-5) is a strong organic acid used as an acid catalyst in organic synthesis and laboratory research. It is available as a solid monohydrate and as a ready-to-use 10 mM solution in DMSO for convenient handling and dosing.
  • Strong organic acid suitable for acid-catalyzed reactions.
  • Ready-to-use 10 mM solution in DMSO for convenient dosing and reduced preparation time.
  • High purity suitable for analytical and synthetic applications.
  • Monohydrate form with defined molecular weight and chemical formula for reproducibility.
  • Applicable to esterification, dehydration, and other acid-catalyzed transformations.
Catalog No. 50-003-03160 Supplier Medchemexpress LLC Supplier No. HYW01517510MM/1ML
p-Toluenesulfonic acid monohydrate (CAS 6192-52-5) is a strong organic acid used as an acid catalyst in organic synthesis and laboratory research. It is available as a solid monohydrate and as a ready-to-use 10 mM solution in DMSO for convenient handling and dosing.
  • Strong organic acid suitable for acid-catalyzed reactions.
  • Ready-to-use 10 mM solution in DMSO for convenient dosing and reduced preparation time.
  • High purity suitable for analytical and synthetic applications.
  • Monohydrate form with defined molecular weight and chemical formula for reproducibility.
  • Applicable to esterification, dehydration, and other acid-catalyzed transformations.