Learn More
Medchemexpress LLC p-Toluenesulfonic acid monohydrate | 6192-52-5 | MFCD00142137 | 99.8% | 190.22 g·mol⁻¹ | C7H10O4S | 500 G
SDP
Catalog No. 5000302522
Encompass_Preferred
p-Toluenesulfonic acid monohydrate is a strong organic sulfonic acid supplied as the monohydrate salt. It is commonly used as an acid catalyst in organic synthesis and for research and analytical applications. The solid material offers high purity and is suitable for laboratory reactions, analytical standards, and catalytic transformations.
  • Strong organic acid and catalyst for dehydration and esterification reactions.
  • Supplied as a monohydrate solid for convenient handling and storage.
  • High purity (≈99.8%) appropriate for research and analytical use.
  • Suitable for use as an analytical standard and reagent in synthesis.
  • Compatible with common polar organic solvents; readily soluble.
  • Available in multiple pack sizes to support small- and large-scale work.
Catalog No. 50-003-02522 Supplier Medchemexpress LLC Supplier No. HYW015175500G
p-Toluenesulfonic acid monohydrate is a strong organic sulfonic acid supplied as the monohydrate salt. It is commonly used as an acid catalyst in organic synthesis and for research and analytical applications. The solid material offers high purity and is suitable for laboratory reactions, analytical standards, and catalytic transformations.
  • Strong organic acid and catalyst for dehydration and esterification reactions.
  • Supplied as a monohydrate solid for convenient handling and storage.
  • High purity (≈99.8%) appropriate for research and analytical use.
  • Suitable for use as an analytical standard and reagent in synthesis.
  • Compatible with common polar organic solvents; readily soluble.
  • Available in multiple pack sizes to support small- and large-scale work.