Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Toluenesulfonic acid monohydrate, ≥99%, MP Biomedicals™

Catalog No. ICN10308790 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10308790 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10308790 Supplier MP Biomedicals, Inc Supplier No. 0210308790
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 6192-52-5
Color White
Density 1.23 to 1.24g/cm3 at 20°C
Quantity 500 g
Physical Form Powder
Assay Percent Range ≥99%
Molecular Formula C7H10O4S
Synonym p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate
InChI Key KJIFKLIQANRMOU-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.O
IUPAC Name 4-methylbenzenesulfonic acid;hydrate
Molecular Weight (g/mol) 190.213
PubChem CID 521998
Formula Weight 190.21
Chemical Name or Material p-Toluenesulfonic acid monohydrate
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.