Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Toluenesulfonic Acid, Monohydrate, Crystal, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02003917 Shop All Avantor Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-003-917 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-003-917 Supplier Avantor Supplier No. W03105

missing translation for 'validMainframeMsg'

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 6192-52-5
Molecular Formula C7H10O4S
Molecular Weight (g/mol) 190.213
InChI Key KJIFKLIQANRMOU-UHFFFAOYSA-N
Synonym p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate
PubChem CID 521998
IUPAC Name 4-methylbenzenesulfonic acid;hydrate
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.O

Specifications

Formula Weight 190.22
Grade BAKER ANALYZED™ Reagent
Quantity 100 g
Flash Point 184.00°C
Packaging Poly Bottle
Physical Form Crystalline
Specific Gravity 1L = 1.23kg
Chemical Name or Material p-Toluenesulfonic Acid, Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.