Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Toluenesulfonic Acid, Monohydrate , MP Biomedical

Catalog No. MP210308710 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210308710 10 g
MP210308780 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP210308710 Supplier MP Biomedicals, Inc Supplier No. 0210308710
Add to Cart
Add to Cart

p-Toluenesulfonic acid monohydrate,>99% purity, is an organic-soluble acid catalyst in organic synthesis. The catalyst has been incorporated in the esterification of carboxylic acids or transesterification of esters.

  • p-Toluenesulfonic acid monohydrate has been used as a reducing agent for the reductive amination of ketones and aldehydes
  • In the presence of p-Toluenesulfonic acid monohydrate novel deazaflavin-cholestane hybrid compounds have been synthesized in a condensation reaction
  • 2-Phenylethyl alpha-glucoside has also been synthesized in the presence of p-Toluenesulfonic acid monohydrate
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-15-4
Molecular Formula C7H8O3S
Molecular Weight (g/mol) 172.20
InChI Key JOXIMZWYDAKGHI-UHFFFAOYSA-N
Synonym p-toluenesulfonic acid, p-toluenesulphonic acid, 4-toluenesulfonic acid, tosic acid, p-tolylsulfonic acid, toluene-4-sulfonic acid, toluenesulfonic acid, benzenesulfonic acid, 4-methyl, p-toluenesulfonate, eltesol
PubChem CID 6101
ChEBI CHEBI:27849

Specifications

Melting Point 97°C to 107°C
Color White
Density 1.23 to 1.24g/cm³ at 20 °C (Lit.)
Packaging Glass
Quantity 10 g
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.