Learn More
Sigma Aldrich P-Toluenesulfonic Acid, Polymer-Bound
Catalog No. 532312100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-231-2100G 100 g
53-231-225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 53-231-2100G Supplier Sigma Aldrich Supplier No. 532312100G
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Scavenges nitrogen nucleophiles, activated ester electrophiles, and bases such as EDC.
Specifications
MDL Number MFCD02683442
Quantity 100 g