Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific p-Toluenesulfonic anhydride, 4124-41-8, MFCD00008548, 25g

Catalog No. 50425650
Encompass_Preferred

Molecular Formula C14H14O5S2, Purity 95+%, Molecular Weight 326.39, Melting Point 121-130°

Catalog No. 50-425-650 Supplier Matrix Scientific Supplier No. 07561525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular Formula C14H14O5S2, Purity 95+%, Molecular Weight 326.39, Melting Point 121-130°

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.