Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Toluenesulfonic anhydride, 97%

Catalog No. aal0016006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0016006 5 g
AAL0016014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0016006 Supplier Thermo Scientific Chemicals Supplier No. L0016006
Only null left
Add to Cart

CAS: 4124-41-8 | C14H14O5S2 | 326.381 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4124-41-8
Molecular Formula C14H14O5S2
Molecular Weight (g/mol) 326.381
MDL Number MFCD00008548
InChI Key PDVFSPNIEOYOQL-UHFFFAOYSA-N
Synonym p-toluenesulfonic anhydride, 4-methylbenzenesulfonic anhydride, 4-toluenesulfonic anhydride, p-toluenesulfonicanhydride, p-toluene sulfonic anhydride, toluene-p-sulphonic anhydride, 4-methylphenyl sulfonyl 4-methylbenzenesulfonate, p-toluenesulphonic anhydride, toluene-4-sulfonic anhydride, benzenesulfonic acid, 4-methyl-, anhydride
PubChem CID 77773
IUPAC Name (4-methylphenyl)sulfonyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OS(=O)(=O)C2=CC=C(C=C2)C

Specifications

Melting Point 125°C to 130°C
Quantity 5 g
UN Number UN3261
Beilstein 2223702
Sensitivity Moisture sensitive
Formula Weight 326.38
Percent Purity 97%
Chemical Name or Material p-Toluenesulfonic anhydride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 223-926-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.