Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich P-Toluenesulfonyl Azide Solution

Catalog No. 71358925G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-358-925G 25 g
71-358-9100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 71-358-925G Supplier Sigma Aldrich Supplier No. 71358925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

p-Toluenesulfonyl azide solution can be employed as a reagent along with alkyne, and amine in the three-component coupling reaction for the synthesis of N-sulfonylamidines in presence of copper catalyst.It can also be used in the synthesis of N-sulfonyl-1,2,3-triazoles by azide-alkyne cycloaddition reaction in the presence of copper catalyst.

Specifications

CAS 941-55-9
Density ∼0.90 g/mL at 20°C
Molecular Formula C7H7N3O2S
Refractive Index n20/D 1.502
Linear Formula C7H7N3O2S
MDL Number MFCD00180767
Quantity 25 g
Synonym 4-Methylbenzenesulfonyl azide solution; p-Toluenesulfonazide solution; Tosyl azide solution
Molecular Weight (g/mol) 197.21 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.