Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich p-Toluenesulfonyl hydrazide

Catalog No. 132004500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-200-4500G 500 g
13-200-4100G 100 g
13-200-425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-200-4500G Supplier Sigma Aldrich Supplier No. 132004500G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

p-Toluenesulfonyl hydrazide

Specifications

CAS 1576-35-8
Melting Point 103°C to 108°C (lit.)
Molecular Formula C7H10N2O2S
Linear Formula CH3C6H4SO2NHNH2
MDL Number MFCD00007588
Quantity 500 g
Synonym p-Toluenesulfonhydrazide; p-Toluenesulfonic acid hydrazide; p-Toluenesulfonyl hydrazide; Tosylhydrazide
Molecular Weight (g/mol) 186.23
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.