Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Toluenesulfonylurea 98.0+%, TCI America™
SDP

Catalog No. T28905G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T28905G Supplier TCI America Supplier No. T28905G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1694-06-0
Molecular Formula C8H10N2O3S
Molecular Weight (g/mol) 214.24
MDL Number MFCD00196522
InChI Key RUTYWCZSEBLPAK-UHFFFAOYSA-N
Synonym n-carbamoyl-4-methylbenzenesulfonamide, 4-toluenesulfonylurea, 4-tolylsulfonylurea, p-toluenesulfonylurea, p-tolylsulfonylurea, n-4-toluenesulfonyl urea, 4-methylbenzene sulfonyl urea, benzenesulfonamide, n-aminocarbonyl-4-methyl, 4-methylbenzenesulfonylurea, amino-n-4-methylphenyl sulfonyl amide
PubChem CID 74327
IUPAC Name (4-methylbenzenesulfonyl)urea
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC(N)=O

Specifications

Melting Point 190°C
Color White
Quantity 5 g
Formula Weight 214.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material p-Toluenesulfonylurea
EINECSNumber (9)-0926
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.