Learn More
Medchemexpress LLC p-Toluic acid (4-Methylbenzoic acid) | 99-94-5 | 99.9% | C8H8O2 | 5 G
SDP
Catalog No. 5000433902
Encompass_Preferred
p-Toluic acid, also known as 4-Methylbenzoic acid, is a substituted benzoic acid and coumarin derivative. This white to off-white solid serves as a synthetic intermediate for compounds such as p-aminomethylbenzoic acid (PAMBA) and p-toluonitrile. It has a molecular weight of 136.15 and a chemical formula of C8H8O2.
  • Functions as an intermediate in the synthesis of other compounds
  • Typically has a purity of 99.93%
  • Soluble in DMSO at 100 mg/mL for in vitro use
  • Intended for research use only
Catalog No. 50-004-33902 Supplier Medchemexpress LLC Supplier No. HY765475G
p-Toluic acid, also known as 4-Methylbenzoic acid, is a substituted benzoic acid and coumarin derivative. This white to off-white solid serves as a synthetic intermediate for compounds such as p-aminomethylbenzoic acid (PAMBA) and p-toluonitrile. It has a molecular weight of 136.15 and a chemical formula of C8H8O2.
  • Functions as an intermediate in the synthesis of other compounds
  • Typically has a purity of 99.93%
  • Soluble in DMSO at 100 mg/mL for in vitro use
  • Intended for research use only