Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC p-Toluic acid | 99-94-5 | 99.9% | C8H8O2 | 25 G
SDP

Catalog No. 5000358681
Encompass_Preferred

p-Toluic acid (4-Methylbenzoic acid) is a substituted benzoic acid and a synthetic p-aminomethylbenzoic acid (PAMBA) intermediate. It is intended for research use only and may have potential reproductive toxicity with repeated high-dose administration.

  • Purity of 99.93%
  • Appears as a white to off-white solid
  • CAS number: 99-94-5
  • Molecular weight: 136.15
  • Chemical formula: C8H8O2
  • Stable at room temperature for 3 years
  • Store in solvent at -80°C for 2 years or -20°C for 1 year
  • Identified uses include laboratory chemicals and manufacturing of substances

Catalog No. 50-003-58681 Supplier Medchemexpress LLC Supplier No. 25GHY76547
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

p-Toluic acid (4-Methylbenzoic acid) is a substituted benzoic acid and a synthetic p-aminomethylbenzoic acid (PAMBA) intermediate. It is intended for research use only and may have potential reproductive toxicity with repeated high-dose administration.

  • Purity of 99.93%
  • Appears as a white to off-white solid
  • CAS number: 99-94-5
  • Molecular weight: 136.15
  • Chemical formula: C8H8O2
  • Stable at room temperature for 3 years
  • Store in solvent at -80°C for 2 years or -20°C for 1 year
  • Identified uses include laboratory chemicals and manufacturing of substances

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.