Learn More
Medchemexpress LLC p-Toluic acid-d3 | 19215-16-8 | >98.0% | 139.17 g/mol | C8H5D3O2 | 10 MG
SDP
Catalog No. 5000298798
Encompass_Preferred
p-Toluic acid-d3 (4-methylbenzoic acid-d3) is the deuterium-labeled form of p-toluic acid, intended for analytical and synthetic use. It is used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, or LC-MS, and as an intermediate in organic synthesis.
  • Used as an internal standard for NMR, GC-MS, and LC-MS quantitative analysis.
  • Suitable as a synthetic intermediate for organic chemistry applications.
  • Available in small-scale quantities for analytical use, e.g., 10 MG.
  • Chemical purity min 98% with isotopic enrichment typically 98 atom % D.
Catalog No. 50-002-98798 Supplier Medchemexpress LLC Supplier No. HY76547S210MG
p-Toluic acid-d3 (4-methylbenzoic acid-d3) is the deuterium-labeled form of p-toluic acid, intended for analytical and synthetic use. It is used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, or LC-MS, and as an intermediate in organic synthesis.
  • Used as an internal standard for NMR, GC-MS, and LC-MS quantitative analysis.
  • Suitable as a synthetic intermediate for organic chemistry applications.
  • Available in small-scale quantities for analytical use, e.g., 10 MG.
  • Chemical purity min 98% with isotopic enrichment typically 98 atom % D.