Learn More
Medchemexpress LLC P-toluic acid-d3 (4-methylbenzoic acid-d3) | 19215-16-8 | 99% | C8H5D3O2 | 25 MG
SDP
Catalog No. 5000359264
Encompass_Preferred
P-Toluic acid-d3 is the deuterium labeled p-Toluic acid. It is a substituted benzoic acid and can be used as an intermediate for the synthesis of para-aminomethylbenzoic acid (PAMBA), p-tolunitrile, and other compounds. Intended for research use only, this compound utilizes stable heavy isotopes for quantitative analysis during drug development, leveraging deuteration's potential to affect pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-59264 Supplier Medchemexpress LLC Supplier No. 25MGHY76547S2
P-Toluic acid-d3 is the deuterium labeled p-Toluic acid. It is a substituted benzoic acid and can be used as an intermediate for the synthesis of para-aminomethylbenzoic acid (PAMBA), p-tolunitrile, and other compounds. Intended for research use only, this compound utilizes stable heavy isotopes for quantitative analysis during drug development, leveraging deuteration's potential to affect pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS