Learn More
Medchemexpress LLC P-Toluic acid-d7 | 1219798-76-1 | 99.7% | C8HD7O2 | 100 MG
SDP
Catalog No. 5000434519
Encompass_Preferred
P-Toluic acid-d7 (4-Methylbenzoic acid-d7) is a deuterium-labeled variant of p-Toluic acid. The unlabeled p-Toluic acid is a substituted benzoic acid and an intermediate in the synthesis of compounds such as para-aminomethylbenzoic acid (PAMBA) and p-tolunitrile. This deuterium-labeled compound is suitable for use as an internal standard in quantitative analytical techniques and as a tracer in various research applications.
  • Deuterium labeled for use as a tracer in research
  • High purity, specified at 99.72%
  • Functions as an internal standard for quantitative analysis, including NMR, GC-MS, or LC-MS
  • Valuable for studying the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-34519 Supplier Medchemexpress LLC Supplier No. HY76547S1100MG
P-Toluic acid-d7 (4-Methylbenzoic acid-d7) is a deuterium-labeled variant of p-Toluic acid. The unlabeled p-Toluic acid is a substituted benzoic acid and an intermediate in the synthesis of compounds such as para-aminomethylbenzoic acid (PAMBA) and p-tolunitrile. This deuterium-labeled compound is suitable for use as an internal standard in quantitative analytical techniques and as a tracer in various research applications.
  • Deuterium labeled for use as a tracer in research
  • High purity, specified at 99.72%
  • Functions as an internal standard for quantitative analysis, including NMR, GC-MS, or LC-MS
  • Valuable for studying the pharmacokinetic and metabolic profiles of drugs