Learn More
Medchemexpress LLC p-Toluic acid-d7 | 1219798-76-1 | MFCD06658857 | 99.72% | C8HD7O2 | 25 MG
SDP
Catalog No. 5000359266
Encompass_Preferred
p-Toluic acid-d7 is the deuterium labeled p-Toluic acid. This substituted benzoic acid can be used as an intermediate for the synthesis of para-aminomethylbenzoic acid (PAMBA) and p-tolunitrile. It is intended for research use only.
  • Deuterium labeled compound
  • Can be used as a tracer
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potential to affect pharmacokinetic and metabolic profiles in drug development
Catalog No. 50-003-59266 Supplier Medchemexpress LLC Supplier No. 25MGHY76547S1
p-Toluic acid-d7 is the deuterium labeled p-Toluic acid. This substituted benzoic acid can be used as an intermediate for the synthesis of para-aminomethylbenzoic acid (PAMBA) and p-tolunitrile. It is intended for research use only.
  • Deuterium labeled compound
  • Can be used as a tracer
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potential to affect pharmacokinetic and metabolic profiles in drug development