Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Toluidine, 99+%

Catalog No. AAA136980I
Change view
Click to view available options
Quantity:
1000 g
5000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA136980I 5000 g
AAA136980B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA136980I Supplier Thermo Scientific Chemicals Supplier No. A136980I
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 106-49-0 | C7H9N | 107.156 g/mol

p-Toluidine is an intermediate in the production of dyes, organic chemicals and aromatic azo compounds. It serves as a component of accelerators for cyanoacrylate glues. Furthermore, it acts as a bidentate Schiff base ligand through condensation with salicylaldehyde. It reacts with catecholamine to form a dye which is useful for spectrophotometric determination of catecholamine drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Toluidine is an intermediate in the production of dyes, organic chemicals and aromatic azo compounds. It serves as a component of accelerators for cyanoacrylate glues. Furthermore, it acts as a bidentate Schiff base ligand through condensation with salicylaldehyde. It reacts with catecholamine to form a dye which is useful for spectrophotometric determination of catecholamine drugs.

Solubility
Soluble in ethanol, pyridine, diethyl ether, acetone, carbon tetrachloride, methanol, carbon disulfide, oils and dilute acids.

Notes
Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-49-0
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00007906
InChI Key RZXMPPFPUUCRFN-UHFFFAOYSA-N
Synonym p-toluidine, 4-toluidine, p-tolylamine, 4-aminotoluene, benzenamine, 4-methyl, 4-methylbenzenamine, p-methylaniline, p-methylbenzenamine, p-toluidin, p-aminotoluene
PubChem CID 7813
ChEBI CHEBI:37825
IUPAC Name 4-methylaniline
SMILES CC1=CC=C(C=C1)N

Specifications

Melting Point 42°C to 46°C
Density 1.05
Boiling Point 199°C to 202°C
Flash Point 88°C (190°F)
Quantity 5000 g
UN Number UN3451
Beilstein 471281
Sensitivity Air and light sensitive
Merck Index 14,9536
Solubility Information Soluble in ethanol,pyridine,diethyl ether,acetone,carbon tetrachloride,methanol,carbon disulfide,oils and dilute acids.
Formula Weight 107.16
Percent Purity ≥99%
Chemical Name or Material p-Toluidine
Show More Show Less
Hazard Category H301+H311+H331-H317-H319-H351
Hazard Statement GHS H Statement
H301-H311-H331-H351-H319-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing cancer.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P312-P330-P333+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOLUIDINES, SOLID
EINECSNumber 203-403-1
RTECSNumber XU3150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.