Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Toluidine, For Spectrophotometric Det. of Au, Tl(III), W, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111018202 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8202 500 g
11-101-9343 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8202 Supplier MilliporeSigma Supelco Supplier No. 89630500G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

MDL Number MFCD00007906
Synonym 4-Aminotoluene; 4-Methylaniline

Specifications

Melting Point 41°C to 46°C; 42°C to 45°C
Density 0.973 g/mL (at 25°C)
CAS 106-49-0
Boiling Point 200°C
Flash Point 87°C - Closed Cup
Ignition Point 899°F
Linear Formula CH3C6H4 NH2
Quantity 500 g
Solubility Information Ethanol: 1 g/10 mL (Hot)
Vapor Density 3.9 (vs. air)
Vapor Pressure 7 hPa (68°C)
Formula Weight 107.15
Percent Purity ≥99.0% (GC); ≥99.0%
Show More Show Less
RTECSNumber XU3150000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.