Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aobchem P-tolyl propionate | 97% | CAS 7495-84-3 | C10H12O2 | 5g

Catalog No. 5001023284
Encompass_Preferred

P-tolyl propionate | 97% | CAS 7495-84-3 | C10H12O2 | 5g

Catalog No. 50-010-23284 Supplier Aobchem Supplier No. A3270755G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

P-tolyl propionate | 97% | CAS 7495-84-3 | C10H12O2 | 5g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.