Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich P-Tolylboronic Acid

Catalog No. 3936221G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-362-21G 1 g
39-362-210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 39-362-21G Supplier Sigma Aldrich Supplier No. 3936221G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

P-Tolylboronic Acid

Specifications

CAS 5720-05-8
Melting Point 256°C to 263°C
Molecular Formula C7H9BO2
Linear Formula CH3C6H4B(OH)2
MDL Number MFCD00039138
Quantity 1 g
Synonym (p-Methylphenyl)boronic acid; 4-Methylbenzeneboronic acid; 4-Methylphenylboronic acid; 4-Tolueneboronic acid; 4-Tolylboronic acid; p-Tolueneboronic acid; NSC 62870; p-Methylbenzeneboronic acid
Molecular Weight (g/mol) 135.96 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.