Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Tolylhydrazine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. T031925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T031925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T031925G Supplier TCI America Supplier No. T031925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 637-60-5
Molecular Formula C7H11ClN2
Quantity 25 g
MDL Number MFCD00012940
UN Number 2811
Synonym p-tolylhydrazine hydrochloride, 4-methylphenylhydrazine hydrochloride, 4-methylphenyl hydrazine hydrochloride, 4-tolylhydrazine hydrochloride, hydrazine, 4-methylphenyl-, monohydrochloride, p-tolylhydrazinium 1+ chloride, hydrazine, p-tolyl-, hydrochloride, 4-methylphenylhydrazine hcl, 1-4-methylphenyl hydrazine hydrochloride, p-tolyl-hydrazine hydrochloride
InChI Key HMHWNJGOHUYVMD-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)NN.Cl
IUPAC Name (4-methylphenyl)hydrazine;hydrochloride
Molecular Weight (g/mol) 158.629
PubChem CID 12504
Formula Weight 158.63
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material p-Tolylhydrazine Hydrochloride
Show More Show Less
EINECSNumber (3)-0468
RTECSNumber MW0195000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.