Learn More
Medchemexpress LLC p-Tolylmaleimide | 1631-28-3 | 98.0% | 100 MG
SDP
Catalog No. 5000341265
Encompass_Preferred
p-Tolylmaleimide (compound 9) is a naphthalimide derivative with cytotoxic effects on cancer cells. It can arrest the cell cycle of human acute myeloid leukemia cells K562 in the sub-G0/G1 phase and induce apoptosis.
  • Inhibits human acute myeloid leukemia cell K562 with IC50s of 24.99 μM (24 h), 6.61 μM (48 h), and 12.10 μM (72 h).
  • Inhibits human acute myeloid leukemia cell Jurkat with IC50s of 27.16 μM (24 h), 15.51 μM (48 h), and 3.94 μM (72 h).
  • Cytotoxicity against human MCF7, NCI-H460, and SF-268 cells (GI50 > 10 μg/mL).
Catalog No. 50-003-41265 Supplier Medchemexpress LLC Supplier No. HY129592100MG
p-Tolylmaleimide (compound 9) is a naphthalimide derivative with cytotoxic effects on cancer cells. It can arrest the cell cycle of human acute myeloid leukemia cells K562 in the sub-G0/G1 phase and induce apoptosis.
  • Inhibits human acute myeloid leukemia cell K562 with IC50s of 24.99 μM (24 h), 6.61 μM (48 h), and 12.10 μM (72 h).
  • Inhibits human acute myeloid leukemia cell Jurkat with IC50s of 27.16 μM (24 h), 15.51 μM (48 h), and 3.94 μM (72 h).
  • Cytotoxicity against human MCF7, NCI-H460, and SF-268 cells (GI50 > 10 μg/mL).