Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC p-Tolylmaleimide | 1631-28-3 | 98.0% | 50 MG
SDP

Catalog No. 5000341274
Encompass_Preferred

p-Tolylmaleimide is a naphthalimide derivative that exhibits cytotoxic effects on cancer cells. It can arrest the cell cycle of human acute myeloid leukemia cells K562 in the sub-G0/G1 phase and induce apoptosis.

  • Cytotoxic effects on cancer cells
  • Arrests cell cycle of human acute myeloid leukemia cells K562
  • Induces apoptosis
  • Solid appearance
  • Light yellow to yellow color
  • Store at 4°C, protect from light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protect from light

Catalog No. 50-003-41274 Supplier Medchemexpress LLC Supplier No. HY12959250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

p-Tolylmaleimide is a naphthalimide derivative that exhibits cytotoxic effects on cancer cells. It can arrest the cell cycle of human acute myeloid leukemia cells K562 in the sub-G0/G1 phase and induce apoptosis.

  • Cytotoxic effects on cancer cells
  • Arrests cell cycle of human acute myeloid leukemia cells K562
  • Induces apoptosis
  • Solid appearance
  • Light yellow to yellow color
  • Store at 4°C, protect from light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protect from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.