Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Tolylurea 98.0+%, TCI America™
SDP

Catalog No. T032825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T032825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T032825G Supplier TCI America Supplier No. T032825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 622-51-5
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.181
MDL Number MFCD00025433
InChI Key DMSHKWHLXNDUST-UHFFFAOYSA-N
Synonym p-tolylurea, p-tolylcarbamide, 4-tolylurea, 4-methylphenyl urea, urea, p-tolyl, 1-p-tolyl urea, 1-4-methylphenyl urea, p-tolycarbamide, urea, 4-methylphenyl, n-4-methylphenyl urea
PubChem CID 12148
IUPAC Name (4-methylphenyl)urea
SMILES CC1=CC=C(C=C1)NC(=O)N

Specifications

Melting Point 180°C
Color White
Quantity 25 g
Formula Weight 150.18
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material p-Tolylurea
RTECSNumber YU4200000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.