Learn More
Uline P-TOUCH D610BTVP LABEL MAKER
Catalog No. NC2649845
Encompass
Portable, battery-powered thermal transfer label printer with QWERTY keyboard, LCD screen, and Bluetooth connectivity for PC label printing software, uses Brother TZe tapes up to 1/2" wide for general office, warehouse, industrial labeling.
Catalog No. NC2649845 Supplier Uline Supplier No. H10159
Portable, battery-powered thermal transfer label printer with QWERTY keyboard, LCD screen, and Bluetooth connectivity for PC label printing software, uses Brother TZe tapes up to 1/2" wide for general office, warehouse, industrial labeling.