Learn More
Medchemexpress LLC p-Tyramine-d4 hydrochloride | 1189884-47-6 | MFCD08704875 | 99.8% | 177.66 | C8H8D4ClNO | 5 MG
SDP
Catalog No. 5000301559
Encompass_Preferred
p-Tyramine-d4 hydrochloride is a deuterium-labeled isotopic analogue of p-tyramine supplied as the hydrochloride salt. It serves as a reference standard and internal standard for quantitative mass spectrometry and isotope-dilution assays in analytical and biochemical research. CAS 1189884-47-6; molecular weight 177.66; molecular formula C8H8D4ClNO.
  • Provides a deuterium-labeled internal standard for accurate quantitation by LC-MS or GC-MS.
  • Suitable for isotope-dilution assays and method development.
  • Supplied as the hydrochloride salt for improved stability and handling.
  • High purity for reliable analytical performance.
  • Available in small pre-weighed quantities for trace-level analyses.
Catalog No. 50-003-01559 Supplier Medchemexpress LLC Supplier No. HYW016823S5MG
p-Tyramine-d4 hydrochloride is a deuterium-labeled isotopic analogue of p-tyramine supplied as the hydrochloride salt. It serves as a reference standard and internal standard for quantitative mass spectrometry and isotope-dilution assays in analytical and biochemical research. CAS 1189884-47-6; molecular weight 177.66; molecular formula C8H8D4ClNO.
  • Provides a deuterium-labeled internal standard for accurate quantitation by LC-MS or GC-MS.
  • Suitable for isotope-dilution assays and method development.
  • Supplied as the hydrochloride salt for improved stability and handling.
  • High purity for reliable analytical performance.
  • Available in small pre-weighed quantities for trace-level analyses.